Assay Details
Target Gene Details
Entrez Gene ID: | 15525 |
Gene Name: | heat shock protein 4 |
Gene Aliases: |
70kDa, AI317151, APG-2, Hsp110, Hsp70RY, mKIAA4025 |
Location: |
Chr.11:53259814-53300479 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 16 - Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hspa4 | NM_008300.3 | NP_032326.3 | ||
| AK053601.1 | 16 | 2271 | ||
| AK054211.1 | 9 | 1059 | ||
| AK133772.1 | ||||
| AK148883.1 | ||||
| AK155305.1 | BAE33178.1 | |||
| AK166102.1 | ||||
| AK220167.1 | BAD90352.1 | |||
| AW545353.1 | ||||
| BC003770.1 | AAH03770.1 | |||
| CA316420.1 | ||||
| D85904.1 | BAA12914.1 |

Genomic Map