Assay Details
Target Gene Details
Entrez Gene ID: | 15525 |
Gene Name: | heat shock protein 4 |
Gene Aliases: |
70kDa, AI317151, APG-2, Hsp110, Hsp70RY, mKIAA4025 |
Location: |
Chr.11:53259814-53300479 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hspa4 | NM_008300.3 | 6 | 922 | NP_032326.3 |
| AK053601.1 | 6 | 856 | ||
| AK054010.1 | 6 | 863 | BAC35620.1 | |
| AK133772.1 | 6 | 864 | ||
| AK155305.1 | 6 | 839 | BAE33178.1 | |
| AK166102.1 | 6 | 907 | ||
| AK179174.1 | 2 | 105 | ||
| AK220167.1 | 6 | 873 | BAD90352.1 | |
| BC003770.1 | 6 | 831 | AAH03770.1 | |
| CF741432.1 | 4 | 341 | ||
| D85904.1 | 6 | 624 | BAA12914.1 |

Genomic Map