Assay Details
Target Gene Details
Entrez Gene ID: | 19014 |
Gene Name: | mediator complex subunit 1 |
Gene Aliases: |
AI480703, CRSP210, DRIP205, PBP, Pparbp, TRAP220, l11Jus15 |
Location: |
Chr.11:98152154-98193314 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Med1 | NM_001080118.1 | 1 | 160 | NP_001073587.1 |
| NM_134027.2 | 1 | 160 | NP_598788.2 | |
| AK005207.1 | 1 | 144 | ||
| AK049203.1 | 1 | 151 | BAC33607.2 | |
| AK054437.1 | 1 | 161 | BAC35779.2 | |
| AK147199.1 | 1 | 137 | BAE27757.1 | |
| BC021440.1 | 1 | 131 | AAH21440.1 | |
| BC079636.1 | 1 | 130 | AAH79636.1 |
More Information
Set Membership: |
|

Genomic Map