Assay Details
Target Gene Details
Entrez Gene ID: | 217026 |
Gene Name: | HEAT repeat containing 6 |
Gene Aliases: |
2700008B19Rik, BB135312, BB249351 |
Location: |
Chr.11:83753397-83783754 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 18 - Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Heatr6 | NM_145432.3 | NP_663407.2 | ||
| AK076219.1 | ||||
| AK132494.1 | ||||
| AK136845.1 | ||||
| AK138219.1 | BAE23587.1 | |||
| BC003942.1 | AAH03942.1 | |||
| BC065092.1 | AAH65092.1 |

Genomic Map