Assay Details
Target Gene Details
Entrez Gene ID: | 217026 |
Gene Name: | HEAT repeat containing 6 |
Gene Aliases: |
2700008B19Rik, BB135312, BB249351 |
Location: |
Chr.11:83753397-83783754 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Heatr6 | NM_145432.3 | 3 | 488 | NP_663407.2 |
| AK012216.1 | 3 | 432 | BAB28103.3 | |
| AK076219.1 | 3 | 616 | ||
| AK132494.1 | 3 | 423 | ||
| AK149059.1 | 3 | 282 | ||
| AK158323.1 | 3 | 618 | ||
| AK169901.1 | 3 | 435 | ||
| BC065092.1 | 3 | 414 | AAH65092.1 | |
| DV657360.1 | 3 | 488 |

Genomic Map