Assay Details
Target Gene Details
Entrez Gene ID: | 16666 |
Gene Name: | keratin 16 |
Gene Aliases: |
AI324768, K16, Krt1-16 |
Location: |
Chr.11:100246091-100248962 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krt16 | NM_008470.1 | NP_032496.1 | ||
| AF053235.1 | AAC79424.1 | |||
| AK137631.1 | BAE23438.1 | |||
| BC103615.1 | AAI03616.1 | |||
| BC103616.1 | AAI03617.1 | |||
| BC103617.1 | AAI03618.1 | |||
| BC103666.1 | AAI03667.1 | |||
| BC114977.1 |

Genomic Map