Assay Details
Target Gene Details
Entrez Gene ID: | 17883 |
Gene Name: | myosin, heavy polypeptide 3, skeletal muscle, embryonic |
Gene Aliases: |
MyHC-emb, Myhs-e, Myhse |
Location: |
Chr.11:67078273-67102291 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Myh3 | NM_001099635.1 | NP_001093105.1 | ||
| AY963798.1 | ||||
| DQ021874.1 | ||||
| DQ021875.1 |

Genomic Map