Assay Details
Target Gene Details
Entrez Gene ID: | 14813 |
Gene Name: | glutamate receptor, ionotropic, NMDA2C (epsilon 3) |
Gene Aliases: |
GluN2C, NMDAR2C, NR2C |
Location: |
Chr.11:115249169-115267297 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Grin2c | NM_010350.2 | NP_034480.2 | ||
| AK147390.1 | ||||
| AK165770.1 | ||||
| BC137849.1 | ||||
| D10694.1 | BAA01536.1 |
More Information
Set Membership: |
|

Genomic Map