Assay Details
Target Gene Details
Entrez Gene ID: | 217026 |
Gene Name: | HEAT repeat containing 6 |
Gene Aliases: |
2700008B19Rik, BB135312, BB249351 |
Location: |
Chr.11:83753397-83783754 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Heatr6 | NM_145432.3 | NP_663407.2 | ||
| AK012216.1 | BAB28103.3 | |||
| AK076219.1 | ||||
| AK132494.1 | ||||
| AK149059.1 | ||||
| AK158323.1 | ||||
| AK169901.1 | ||||
| BC065092.1 | AAH65092.1 | |||
| DV657360.1 |

Genomic Map