Assay Details
Target Gene Details
Entrez Gene ID: | 268481 |
Gene Name: | keratin 222 |
Gene Aliases: |
6330509G02Rik |
Location: |
Chr.11:99232761-99244281 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krt222 | NM_172946.2 | 3 | 409 | NP_766534.1 |
| AK031954.1 | 3 | 410 | BAC27620.1 | |
| AK044068.1 | 3 | 509 | ||
| AK138778.1 | 3 | 465 | BAE23776.1 | |
| BC079890.1 | 3 | 403 | AAH79890.1 |

Genomic Map