Assay Details
Target Gene Details
Entrez Gene ID: | 268481 |
Gene Name: | keratin 222 |
Gene Aliases: |
6330509G02Rik |
Location: |
Chr.11:99232761-99244281 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krt222 | NM_172946.2 | NP_766534.1 | ||
| AK031954.1 | BAC27620.1 | |||
| AK044068.1 | ||||
| AK138778.1 | BAE23776.1 | |||
| BC079890.1 | AAH79890.1 |

Genomic Map