Assay Details
Target Gene Details
Entrez Gene ID: | 108072 |
Gene Name: | glutamate receptor, metabotropic 6 |
Gene Aliases: |
BC021919, Gm3, Gprc1f, mGluR6, nerg1, nob2, nob3, nob4 |
Location: |
Chr.11:50850008-50866208 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Grm6 | NM_173372.2 | NP_775548.2 | ||
| BC041684.1 | AAH41684.1 | |||
| BC152480.1 |
More Information
Set Membership: |
|

Genomic Map