Assay Details
Target Gene Details
Entrez Gene ID: | 380728 |
Gene Name: | potassium voltage-gated channel, subfamily H (eag-related), member 4 |
Gene Aliases: |
BEC2 |
Location: |
Chr.11:100740378-100760199 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kcnh4 | NM_001081194.2 | NP_001074663.1 | ||
| BC150990.1 | ||||
| BC150994.1 |
More Information
Set Membership: |
|

Genomic Map