Assay Details
Target Gene Details
Entrez Gene ID: | 75706 |
Gene Name: | keratin 24 |
Gene Aliases: |
2310058N18Rik, 2310075C18Rik |
Location: |
Chr.11:99280093-99285238 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krt24 | NM_029393.1 | 8 | 1620 | NP_083669.1 |
| AK009986.1 | 7 | 965 | BAB26628.1 | |
| AK010165.1 | ||||
| AK053678.1 | BAC35470.1 | |||
| BC129847.1 | 8 | 1620 | ||
| BC129848.1 | 8 | 1620 |

Genomic Map