Assay Details
Target Gene Details
Entrez Gene ID: | 26398 |
Gene Name: | mitogen-activated protein kinase kinase 4 |
Gene Aliases: |
JNKK1, MEK4, MKK4, PRKMK4, Sek1, Serk1 |
Location: |
Chr.11:65688244-65788354 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Map2k4 | NM_009157.4 | NP_033183.1 | ||
| AK036928.1 | BAC29642.1 | |||
| AK044534.1 | BAC31968.1 | |||
| AK044698.1 | BAC32038.1 | |||
| AK139777.1 | ||||
| BC029833.1 | AAH29833.1 | |||
| U18310.1 | AAB81554.1 |
More Information
Set Membership: |
|

Genomic Map