Assay Details
Target Gene Details
Entrez Gene ID: | 320864 |
Gene Name: | keratin 26 |
Gene Aliases: |
4732407F15Rik, Krt25b |
Location: |
Chr.11:99328484-99338286 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krt26 | NM_001033397.5 | NP_001028569.2 | ||
| AK028591.1 | ||||
| AK028682.1 | ||||
| AK132445.1 | BAE21173.1 | |||
| AK162718.1 | BAE37035.1 | |||
| BC116672.1 |

Genomic Map