Assay Details
Target Gene Details
Entrez Gene ID: | 16675 |
Gene Name: | keratin 27 |
Gene Aliases: |
Krt1-c29, mIRSa3.1 |
Location: |
Chr.11:99345566-99351094 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krt27 | NM_010666.1 | NP_034796.1 | ||
| AB013607.1 | BAA34228.1 | |||
| AK077384.1 | ||||
| BC140996.1 | ||||
| BC140997.1 |

Genomic Map