Assay Details
Target Gene Details
Entrez Gene ID: | 26399 |
Gene Name: | mitogen-activated protein kinase kinase 6 |
Gene Aliases: |
MEK6, MKK6, Prkmk6, SAPKK3 |
Location: |
Chr.11:110399102-110516212 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Map2k6 | NM_011943.2 | NP_036073.1 | ||
| AK043482.1 | BAC31556.1 | |||
| AK077712.1 | BAC36972.1 | |||
| AK138757.1 | BAE23771.1 | |||
| AK140289.1 | ||||
| AK141708.1 | BAE24808.1 | |||
| AK162671.1 | BAE37014.1 | |||
| BC075652.1 | AAH75652.1 | |||
| U39066.1 | AAB03709.1 | |||
| X97052.1 | CAA65764.1 | |||
| X98067.1 | CAA66670.1 |
More Information
Set Membership: |
|

Genomic Map