Assay Details
Target Gene Details
Entrez Gene ID: | 16660 |
Gene Name: | keratin 31 |
Gene Aliases: |
Ha1, Kha1, Krt1-1, MKHA-1 |
Location: |
Chr.11:100046646-100050551 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krt31 | NM_010659.2 | NP_034789.2 | ||
| M27734.1 | AAA39372.1 |

Genomic Map