Assay Details
Target Gene Details
Entrez Gene ID: | 16670 |
Gene Name: | keratin 32 |
Gene Aliases: |
Ha3, Hka2, K32, Krt1-2, Krtha2 |
Location: |
Chr.11:100080848-100088226 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krt32 | NM_001159374.2 | NP_001152846.2 | ||
| AK009595.1 | ||||
| BC117553.1 | ||||
| BC118064.1 | ||||
| X75649.1 | CAA53304.1 |

Genomic Map