Assay Details
Target Gene Details
Entrez Gene ID: | 53859 |
Gene Name: | mitogen-activated protein kinase kinase kinase 14 |
Gene Aliases: |
Nik, aly |
Location: |
Chr.11:103219764-103267401 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Map3k14 | NM_016896.3 | 14 | 2574 | NP_058592.1 |
| AF143094.1 | 13 | 2491 | AAD31512.1 | |
| AK036200.1 | 14 | 2575 | BAC29344.1 | |
| AK172518.1 | 14 | 2545 | ||
| BC107015.2 | AAI07016.1 |
More Information
Set Membership: |
|

Genomic Map