Assay Details
Target Gene Details
Entrez Gene ID: | 67493 |
Gene Name: | methyltransferase like 16 |
Gene Aliases: |
2610100D03Rik, 2810013M15Rik, A830095F14Rik, AI846133, Mett10d |
Location: |
Chr.11:74770836-74818694 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mettl16 | NM_026197.3 | NP_080473.1 | ||
| AK011780.1 | BAB27835.1 | |||
| AK012739.1 | BAB28440.1 | |||
| AK013306.1 | ||||
| AK029045.1 | ||||
| AK145022.1 | BAE26189.1 | |||
| AK145662.1 | BAE26574.1 | |||
| AK168798.1 | BAE40630.1 | |||
| BC049897.1 | AAH49897.1 | |||
| BC054724.1 | AAH54724.1 |
More Information
Set Membership: |
|

Genomic Map