Assay Details
Target Gene Details
Entrez Gene ID: | 15953 |
Gene Name: | interferon gamma inducible protein 47 |
Gene Aliases: |
47kDa, IRG-47, Ifggc1, Igrd, Iigp4, Iipg4, Irgd |
Location: |
Chr.11:49076587-49096987 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ifi47 | NM_001271676.1 | 1 | 75 | NP_001258605.1 |
| NM_001271677.1 | 2 | 134 | NP_001258606.1 | |
| NM_008330.2 | 1 | 75 | NP_032356.1 | |
| AK156785.1 | BAE33853.1 | |||
| BC001986.1 | AAH01986.1 | |||
| BC009131.1 | AAH09131.1 | |||
| BU696734.1 | 2 | 134 | ||
| BY159742.1 | 1 | 76 | ||
| M63630.1 | AAA37755.1 |
Target Gene Details
Entrez Gene ID: | 18356 |
Gene Name: | olfactory receptor 56 |
Gene Aliases: |
IF7, MOR276-1 |
Location: |
Chr.11:49050733-49135387 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Olfr56 | NM_010999.2 | 2 | 160 | NP_035129.1 |
| AK053352.1 | 2 | 160 | BAC35358.1 | |
| AK133471.1 | 1 | 59 | ||
| BC141219.1 | ||||
| BC147437.1 |
More Information
Set Membership: |
|

Genomic Map