Assay Details
Target Gene Details
Entrez Gene ID: | 68671 |
Gene Name: | phosphate cytidylyltransferase 2, ethanolamine |
Gene Aliases: |
1110033E03Rik, ET |
Location: |
Chr.11:120610086-120617949 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pcyt2 | NM_024229.2 | NP_077191.2 | ||
| AK140458.1 | BAE24398.1 | |||
| AK144544.1 | ||||
| AK166319.1 | BAE38702.1 | |||
| AK199974.1 | ||||
| BC003473.1 | AAH03473.1 | |||
| BC008276.1 | AAH08276.1 |
More Information
Set Membership: |
|

Genomic Map