Assay Details
Target Gene Details
Entrez Gene ID: | 69723 |
Gene Name: | RPA interacting protein |
Gene Aliases: |
2400006N03Rik, Rip |
Location: |
Chr.11:70970200-70977933 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rpain | NM_001252413.1 | NP_001239342.1 | ||
| NM_001252414.1 | NP_001239343.1 | |||
| NM_001252415.1 | NP_001239344.1 | |||
| NM_027186.2 | NP_081462.1 | |||
| AK010289.1 | BAB26825.1 | |||
| BC047996.1 | AAH47996.1 | |||
| CB588111.1 | ||||
| DQ810267.1 | ||||
| DQ810268.1 | ||||
| DQ810269.1 | ||||
| DQ810270.1 | ||||
| DQ810271.1 | ||||
| DQ810272.1 |

Genomic Map