Assay Details
Target Gene Details
Entrez Gene ID: | 237868 |
Gene Name: | sterile alpha and HEAT/Armadillo motif containing 1 |
Gene Aliases: |
A830091I15Rik, C78606, MyD885, Sarm |
Location: |
Chr.11:78472330-78497820 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sarm1 | NM_001168521.1 | 3 | 1499 | NP_001161993.1 |
| NM_172795.3 | 3 | 1499 | NP_766383.2 | |
| AK172964.1 | 3 | 1502 | BAD32242.1 | |
| AY444167.1 | 3 | 1203 | AAR17521.1 | |
| BC052098.1 | 3 | 1145 | ||
| BC058534.1 | 3 | 1446 | AAH58534.1 | |
| BC080850.1 | 3 | 1499 | AAH80850.1 |
More Information
Set Membership: |
|

Genomic Map