Assay Details
Target Gene Details
Entrez Gene ID: | 193116 |
Gene Name: | SLU7 splicing factor homolog (S. cerevisiae) |
Gene Aliases: |
AU018913, D11Ertd730e, D3Bwg0878e |
Location: |
Chr.11:43433723-43447982 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slu7 | NM_148673.3 | 12 | 1239 | NP_683514.2 |
| NM_198936.1 | 11 | 1160 | NP_945174.1 | |
| AK029117.1 | 11 | 1130 | BAC26306.1 | |
| AK046262.1 | 11 | 1131 | BAC32662.1 | |
| AK047597.1 | 12 | 1227 | BAC33093.1 | |
| AK049178.1 | 11 | 1129 | BAC33589.1 | |
| BC013810.1 | 9 | 759 | AAH13810.1 | |
| BC023057.1 | AAH23057.1 | |||
| BC060954.1 | 11 | 1128 | AAH60954.1 | |
| BC082780.1 | 12 | 1219 | AAH82780.1 | |
| BC106099.1 | 11 | 1141 | AAI06100.1 |
More Information
Set Membership: |
|

Genomic Map