Assay Details
Target Gene Details
Entrez Gene ID: | 192159 |
Gene Name: | pre-mRNA processing factor 8 |
Gene Aliases: |
AU019467, D11Bwg0410e, DBF3/PRP8, Prp8, Sfprp8l |
Location: |
Chr.11:75486777-75509447 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prpf8 | NM_138659.2 | 16 | 2786 | NP_619600.2 |
| AB047391.1 | 16 | 2512 | BAB32671.1 | |
| AK144232.1 | 15 | 2546 | BAE25786.1 | |
| BC093481.1 | 16 | 2716 |
More Information
Set Membership: |
|

Genomic Map