Assay Details
Target Gene Details
Entrez Gene ID: | 19921 |
Gene Name: | ribosomal protein L19 |
Gene Aliases: |
- |
Location: |
Chr.11:98023080-98030493 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rpl19 | NM_001159483.1 | NP_001152955.1 | ||
| CB203522.1 |
More Information
Set Membership: |
|

Genomic Map