Assay Details
Target Gene Details
Entrez Gene ID: | 192786 |
Gene Name: | Rap guanine nucleotide exchange factor (GEF) 6 |
Gene Aliases: |
A530068K01, AI844632, BB085664, C030018K18Rik, PDZ-GEF2, Pdzgef2, RA-GEF-2, Ragef2, mKIAA4052 |
Location: |
Chr.11:54521754-54699286 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rapgef6 | NM_001252494.1 | 8 | 964 | NP_001239423.1 |
| NM_001252496.1 | 8 | 964 | NP_001239425.1 | |
| NM_001252497.1 | 3 | 1222 | NP_001239426.1 | |
| NM_001252498.1 | 3 | 1222 | NP_001239427.1 | |
| NM_175258.4 | 8 | 964 | NP_780467.2 | |
| AK045540.1 | 8 | 906 | BAC32411.1 | |
| AK049228.1 | 8 | 920 | BAC33622.1 | |
| AK049669.1 | 3 | 1223 | ||
| AK164343.1 | 3 | 1195 | BAE37750.1 | |
| AK167055.1 | 8 | 965 | BAE39219.1 | |
| BC141182.1 | 8 | 863 |
More Information
Set Membership: |
|

Genomic Map