Assay Details
Target Gene Details
Entrez Gene ID: | 19921 |
Gene Name: | ribosomal protein L19 |
Gene Aliases: |
- |
Location: |
Chr.11:98023080-98030493 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rpl19 | NM_001159483.1 | NP_001152955.1 | ||
| NM_009078.2 | NP_033104.2 | |||
| AK010440.1 | BAB26941.1 | |||
| AK075817.1 | ||||
| AK168232.1 | ||||
| AK168481.1 | BAE40369.1 | |||
| BC087961.1 | AAH87961.1 | |||
| BC089549.1 | AAH89549.1 | |||
| BC092539.1 | AAH92539.1 | |||
| BU524941.1 | ||||
| CB203522.1 |
More Information
Set Membership: |
|

Genomic Map