Assay Details
Target Gene Details
Entrez Gene ID: | 268449 |
Gene Name: | ribosomal protein L23A |
Gene Aliases: |
BC029892, MDA20 |
Location: |
Chr.11:78180935-78183584 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rpl23a | NM_207523.2 | NP_997406.1 | ||
| AF452721.1 | AAQ04686.1 | |||
| AK028387.1 | BAE20441.1 | |||
| AK152095.1 | BAE30943.1 | |||
| BC016558.1 | AAH16558.1 | |||
| BC026656.1 | AAH26656.1 | |||
| BC029892.1 | AAH29892.1 | |||
| BC086883.1 | AAH86883.1 | |||
| BC086884.1 | AAH86884.1 | |||
| BC096637.1 | AAH96637.1 | |||
| BC125640.1 | ||||
| BC125642.1 | ||||
| BY271596.1 |
More Information
Set Membership: |
|

Genomic Map