Assay Details
Target Gene Details
Entrez Gene ID: | 74637 |
Gene Name: | sedoheptulokinase |
Gene Aliases: |
4930431K22Rik, AI194947, AW260459, Carkl |
Location: |
Chr.11:73199224-73224506 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Shpk | NM_029031.3 | NP_083307.1 | ||
| AK015273.1 | BAB29776.1 | |||
| AK171027.1 | ||||
| BC054721.1 | AAH54721.1 | |||
| BC056433.1 | AAH56433.1 |
More Information
Set Membership: |
|

Genomic Map