Assay Details
Target Gene Details
Entrez Gene ID: | 268480 |
Gene Name: | Rap guanine nucleotide exchange factor (GEF)-like 1 |
Gene Aliases: |
B230105J10 |
Location: |
Chr.11:98836785-98853006 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rapgefl1 | NM_001080925.1 | NP_001074394.1 | ||
| BC080279.1 | AAH80279.1 | |||
| BC130266.1 |
More Information
Set Membership: |
|

Genomic Map