Assay Details
Target Gene Details
Entrez Gene ID: | 59040 |
Gene Name: | ras homolog gene family, member T1 |
Gene Aliases: |
2210403N23Rik, AA415293, AF244542, AI834919, Arht1, C430039G08Rik, Miro1 |
Location: |
Chr.11:80209005-80267907 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rhot1 | NM_001163354.1 | NP_001156826.1 | ||
| NM_001163355.1 | NP_001156827.1 | |||
| NM_021536.7 | NP_067511.4 | |||
| AK019059.1 | BAB31529.2 | |||
| AK028597.1 | BAC26025.1 | |||
| AK031139.1 | BAC27274.1 | |||
| AK032096.1 | ||||
| AK041119.1 | BAC30828.1 | |||
| AK043323.1 | ||||
| AK053263.1 | ||||
| AK054472.1 | BAC35792.1 | |||
| AK078955.1 | BAC37478.1 | |||
| AK090071.1 | ||||
| AK138830.1 | BAE23792.1 | |||
| AK141621.1 | BAE24771.1 | |||
| AK147749.1 | BAE28113.1 | |||
| AK147801.1 | BAE28148.1 | |||
| AK167502.1 | BAE39578.1 | |||
| AK169811.1 | ||||
| AK191854.1 | ||||
| BC046785.1 | AAH46785.1 | |||
| BC058350.1 | AAH58350.1 |
More Information
Set Membership: |
|

Genomic Map