Assay Details
Target Gene Details
Entrez Gene ID: | 209011 |
Gene Name: | sirtuin 7 |
Gene Aliases: |
- |
Location: |
Chr.11:120618372-120625002 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sirt7 | NM_153056.2 | NP_694696.2 | ||
| AK135484.1 | ||||
| AY251540.1 | AAP83960.1 | |||
| BC026403.1 | AAH26403.1 | |||
| BC026650.1 | AAH26650.1 | |||
| BC040759.1 |
More Information
Set Membership: |
|

Genomic Map