Assay Details
Target Gene Details
Entrez Gene ID: | 18082 |
Gene Name: | 4-nitrophenylphosphatase domain and non-neuronal SNAP25-like protein homolog 1 (C. elegans) |
Gene Aliases: |
- |
Location: |
Chr.11:4873965-4894202 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nipsnap1 | NM_008698.2 | NP_032724.1 | ||
| AJ001260.1 | CAA04634.1 | |||
| AK013140.1 | ||||
| AK031804.1 | BAC27558.1 | |||
| AK147786.1 | BAE28136.1 | |||
| BC010837.1 | AAH10837.1 |

Genomic Map