Assay Details
Target Gene Details
Entrez Gene ID: | 19401 |
Gene Name: | retinoic acid receptor, alpha |
Gene Aliases: |
Nr1b1, RAR, RARalpha1 |
Location: |
Chr.11:98927809-98974942 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rara | NM_001176528.1 | 2 | 736 | NP_001169999.1 |
| NM_001177302.1 | 4 | 941 | NP_001170773.1 | |
| NM_001177303.1 | 3 | 686 | NP_001170774.1 | |
| NM_009024.2 | 3 | 808 | NP_033050.2 | |
| AB221572.1 | 2 | 255 | ||
| AK042163.1 | 3 | 687 | BAE20623.1 | |
| AK153644.1 | 2 | 282 | BAE32132.1 | |
| AK154623.1 | 3 | 781 | BAE32722.1 | |
| AK170956.1 | 2 | 736 | ||
| BC010216.1 | 3 | 806 | AAH10216.1 | |
| BC090396.1 | 2 | 352 | AAH90396.1 | |
| CT010231.1 | 2 | 251 | CAJ18439.1 | |
| S56656.1 | 3 | 788 | AAB25783.1 | |
| X56565.1 | CAA39917.1 | |||
| X56566.1 | ||||
| X56567.1 | ||||
| X56568.1 | ||||
| X56570.1 | ||||
| X56571.1 | ||||
| X56572.1 | CAA39919.1 |
More Information
Set Membership: |
|

Genomic Map