Assay Details
Target Gene Details
Entrez Gene ID: | 52892 |
Gene Name: | SCO cytochrome oxidase deficient homolog 1 (yeast) |
Gene Aliases: |
2610001C07Rik, D11Bwg1310e |
Location: |
Chr.11:67052670-67067444 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sco1 | NM_001040026.1 | NP_001035115.1 | ||
| AK011251.1 | ||||
| AK017794.1 | ||||
| AK139512.1 | BAE24044.1 | |||
| AK167305.1 | BAE39409.1 | |||
| BC139008.1 | ||||
| BC139009.1 |
More Information
Set Membership: |
|

Genomic Map