Assay Details
Target Gene Details
Entrez Gene ID: | 21402 |
Gene Name: | S-phase kinase-associated protein 1A |
Gene Aliases: |
15kDa, 2610043E24Rik, 2610206H23Rik, EMC19, OCP-II, OCP2, SKP1, Tceb1l, p19A, p19Skp1 |
Location: |
Chr.11:52230522-52246858 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Skp1a | NM_011543.4 | NP_035673.3 | ||
| AF083214.1 | AAD16036.1 | |||
| AK002983.1 | BAB22496.1 | |||
| AK010628.1 | BAB27074.1 | |||
| AK012498.1 | BAB28281.1 | |||
| AK014245.1 | BAB29222.1 | |||
| AK027909.1 | BAC25660.1 | |||
| AK078330.1 | BAC37220.1 | |||
| AK088339.1 | BAC40292.1 | |||
| AK152609.1 | BAE31355.1 | |||
| AK160198.1 | BAE35688.1 | |||
| AK166673.1 | BAE38934.1 | |||
| BC002115.1 | AAH02115.1 | |||
| BU841654.1 | ||||
| Z47088.1 |
More Information
Set Membership: |
|

Genomic Map