Assay Details
Target Gene Details
Entrez Gene ID: | 217011 |
Gene Name: | notchless homolog 1 (Drosophila) |
Gene Aliases: |
AL022765, BC018399, Nle, l11Jus1, l11Jus4 |
Location: |
Chr.11:82900768-82908445 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nle1 | NM_145431.2 | NP_663406.2 | ||
| AK141230.1 | BAE24602.1 | |||
| AK170853.1 | BAE42074.1 | |||
| BC018399.1 | AAH18399.1 |

Genomic Map