Assay Details
Target Gene Details
Entrez Gene ID: | 268451 |
Gene Name: | RAB11 family interacting protein 4 (class II) |
Gene Aliases: |
A730072L08Rik, RAB11-FIP4, mKIAA1821 |
Location: |
Chr.11:79591021-79698023 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rab11fip4 | NM_175543.3 | NP_780752.1 | ||
| AK046734.1 | BAC32850.1 | |||
| AK147219.1 | ||||
| AK147344.1 |
Target Gene Details
Entrez Gene ID: | 790912 |
Gene Name: | RAB11 family interacting protein 4 (class II), opposite strand 1 |
Gene Aliases: |
Gm11202, OTTMUSG00000000185 |
Location: |
Chr.11:79607079-79623163 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rab11fip4os1 | NR_003283.1 | |||
| AK049292.1 |
More Information
Set Membership: |
|

Genomic Map