Assay Details
Target Gene Details
Entrez Gene ID: | 20500 |
Gene Name: | solute carrier family 13 (sodium-dependent dicarboxylate transporter), member 2 |
Gene Aliases: |
Nadc1, mNaDC-1 |
Location: |
Chr.11:78397276-78422280 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc13a2 | NM_022411.3 | NP_071856.1 | ||
| AF201903.1 | AAG15426.1 | |||
| AK140972.1 | ||||
| BC013493.1 | AAH13493.1 |
More Information
Set Membership: |
|

Genomic Map