Assay Details
Target Gene Details
Entrez Gene ID: | 19324 |
Gene Name: | RAB1, member RAS oncogene family |
Gene Aliases: |
Gtbp, Rab-1, Rab1A, Ypt1, mKIAA3012 |
Location: |
Chr.11:20201602-20226856 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rab1 | NM_008996.3 | NP_033022.1 | ||
| AB232583.1 | BAF02845.1 | |||
| AF226873.1 | AAF33844.1 | |||
| AK034408.1 | BAC28697.1 | |||
| AK050526.1 | 3 | 372 | ||
| AK129477.1 | BAC98287.1 | |||
| AK151085.1 | BAE30098.1 | |||
| AK152978.1 | ||||
| AK159425.1 | BAE35072.1 | |||
| AK166249.1 | BAE38660.1 | |||
| AK167150.1 | BAE39292.1 | |||
| BC002077.1 | AAH02077.3 | |||
| Y00094.1 | CAA68284.1 |

Genomic Map