Assay Details
Target Gene Details
Entrez Gene ID: | 19376 |
Gene Name: | RAB34, member RAS oncogene family |
Gene Aliases: |
AI326479, Narr, Rah, Rah1 |
Location: |
Chr.11:78187879-78192193 on Build GRCm38 |
Assay Gene Location: | Overlaps - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rab34 | NM_001159482.1 | NP_001152954.1 | ||
| AA041780.1 | ||||
| AF327929.1 | AAK11241.1 | |||
| BY317641.1 |

Genomic Map