Assay Details
Target Gene Details
Entrez Gene ID: | 20370 |
Gene Name: | seizure related gene 6 |
Gene Aliases: |
BSRP-C, D11Bhm177e, sez-6 |
Location: |
Chr.11:77930839-77979052 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sez6 | NM_001291225.1 | 4 | 1168 | NP_001278154.1 |
| NM_021286.4 | 4 | 1168 | NP_067261.2 | |
| AB206791.1 | 4 | 1104 | BAE44444.1 | |
| AK032277.1 | 4 | 1092 | ||
| AK147705.1 | 4 | 1087 | ||
| AK165757.1 | 4 | 952 | ||
| AK220513.1 | 3 | 1798 | BAD90303.1 | |
| BC053011.1 | 4 | 1064 | AAH53011.1 | |
| BC055345.1 | 4 | 1168 | AAH55345.1 | |
| D29763.1 | 4 | 995 | BAA06167.1 | |
| D64009.1 | 4 | 968 | BAA10889.1 | |
| D64010.1 | 4 | 968 | BAA10890.1 | |
| X81719.1 | 1 | 98 |

Genomic Map