Assay Details
Target Gene Details
Entrez Gene ID: | 237831 |
Gene Name: | solute carrier family 13 (sodium-dependent citrate transporter), member 5 |
Gene Aliases: |
Indy, NaC2/NaCT, Nact, mINDY |
Location: |
Chr.11:72241990-72267032 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc13a5 | NM_001004148.4 | NP_001004148.1 | ||
| AK043499.1 | BAE20640.1 | |||
| AK048662.1 | BAE20664.1 | |||
| AK048944.1 | ||||
| AK140943.1 | ||||
| AK158253.1 | ||||
| AY368893.1 | AAR14317.1 |
More Information
Set Membership: |
|

Genomic Map