Assay Details
Target Gene Details
Entrez Gene ID: | 637515 |
Gene Name: | NLR family, pyrin domain containing 1B |
Gene Aliases: |
ENSMUSG00000070390, Nalp1b, OTTMUSG00000006087, OTTMUSG00000006089 |
Location: |
Chr.11:71153014-71230774 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nlrp1b | NM_001040696.1 | NP_001035786.1 | ||
| DQ117591.1 | AAZ40517.1 | |||
| DQ153214.1 | AAZ40529.1 |
More Information
Set Membership: |
|

Genomic Map