Assay Details
Target Gene Details
Entrez Gene ID: | 18208 |
Gene Name: | netrin 1 |
Gene Aliases: |
AI561871, BC019633, Netrin-1 |
Location: |
Chr.11:68209364-68386826 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ntn1 | NM_008744.2 | NP_032770.2 | ||
| AF128865.1 | AAD28602.1 | |||
| AI840271.1 | ||||
| BC141294.1 | ||||
| BE652257.1 | ||||
| U65418.1 | AAC52971.1 |
More Information
Set Membership: |
|

Genomic Map