Assay Details
Target Gene Details
Entrez Gene ID: | 19376 |
Gene Name: | RAB34, member RAS oncogene family |
Gene Aliases: |
AI326479, Narr, Rah, Rah1 |
Location: |
Chr.11:78187879-78192193 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rab34 | NM_001159482.1 | 2 | 234 | NP_001152954.1 |
| NM_033475.3 | 1 | 579 | NP_258436.2 | |
| AB082927.1 | 1 | 493 | BAC16803.1 | |
| AB232630.1 | BAF02892.1 | |||
| AF322068.1 | AAK09398.1 | |||
| AF327929.1 | 2 | 187 | AAK11241.1 | |
| BB643883.1 | 1 | 68 | ||
| BC038638.1 | 1 | 54 | AAH38638.1 | |
| BY317641.1 | 2 | 224 |

Genomic Map